missing translation for 'onlineSavingsMsg'
Learn More

DL-Homoserine lactone hydrobromide, MP Biomedicals™

Product Code. 11413872 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413872 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413872 Supplier MP Biomedicals Supplier No. 0215738905
  • ONLINE EXCLUSIVE PRICE
    111.00 EUR
    105.00 EUR / 5g
    Save 6.00 EUR
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 6305-38-0
Color White
Molecular Formula C4H8BrNO2
Quantity 5 g
Synonym alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
InChI Key MKLNTBLOABOJFZ-UHFFFAOYSA-N
SMILES C1COC(=O)C1N.Br
IUPAC Name 3-aminooxolan-2-one;hydrobromide
Molecular Weight (g/mol) 182.017
PubChem CID 73508
Formula Weight 182.02
Physical Form Powder
Chemical Name or Material DL-Homoserine lactone hydrobromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.