CAS RN 626-95-9
CAS RN 626-95-9
IUPAC Name:
pentane-1,4-diol
Synonyms:
1,4-dihydroxypentane
Molecular Weight (g/mol):
104.15
Molecular Formula:
C5H12O2
InChi Key:
GLOBUAZSRIOKLN-UHFFFAOYNA-N
SMILES:
CC(O)CCCO
1
–
2
of
2
results
1
1,4-Pentanediol, TRC
CAS: 626-95-9 Molecular Formula: C5 H12 O2 Molecular Weight (g/mol): 104.148 Synonym: 1,4-Dihydroxy-1-methylbutane,1,4-Dihydroxypentane,2,5-Pentanediol,4-Hydroxy-1-pentanol IUPAC Name: pentane-1,4-diol SMILES: CC(O)CCCO