Learn More
1,4-Pentanediol, TRC
Product Code. 17979734
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979734 5 g 5g
17958664 1 g 1g
17929444 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17979734 Supplier Toronto Research Chemicals Supplier No. TRCP2734005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 626-95-9
Molecular Formula C5 H12 O2
Molecular Weight (g/mol) 104.148
Synonym 1,4-Dihydroxy-1-methylbutane, 1,4-Dihydroxypentane, 2,5-Pentanediol, 4-Hydroxy-1-pentanol
IUPAC Name pentane-1,4-diol
SMILES CC(O)CCCO
Specifications
Quantity 5 g
InChI Formula InChI=1S/C5H12O2/c1-5(7)3-2-4-6/h5-7H,2-4H2,1H3
Formula Weight 104.0837
Chemical Name or Material 1,4-Pentanediol