accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 620-02-0

IUPAC Name: 5-methylfuran-2-carbaldehyde
Synonyms: 5-methylfurfural 5-methyl-2-furfuraldehyde 5-methyl-2-furfural
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: OUDFNZMQXZILJD-UHFFFAOYSA-N
SMILES: CC1=CC=C(O1)C=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)