accessibility menu, dialog, popup

UserName

NH2

CAS RN 62-53-3

IUPAC Name: aniline
Synonyms: aniline epa pesticide chemical code 251400 aromatic amine benzene,amino (aniline) 2-(13)C-labeled cpd aniline (14)C-labeled cpd aniline anyvim aniline, 3-(13)C-labeled cpd aniline, magnesium (1:1) salt conjugate acid aniline
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: PAYRUJLWNCNPSJ-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1
Quantity 
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (12)
  • (6)
  • (3)
Grade 
  • (3)
  • (4)
  • (1)
  • (4)
  • (4)
  • (3)
Percent Purity 
  • (4)
  • (3)
  • (4)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (12)
  • (6)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (12)
  • (6)
  • (3)
  • (3)
  • (4)
  • (1)
  • (4)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (8)
  • (2)
  • (3)
  • (2)
  • (2)