Learn More
Aniline, Supelco™
Description
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
Specifications
Specifications
| CAS | 62-53-3 |
| Melting Point | -6.2°C |
| Density | 1.02 g/cm3 at 20°C |
| Boiling Point | 184°C/1013 hPa |
| MDL Number | MFCD00007629 |
| Quantity | 1 L |
| Synonym | Aniline, Aminobenzene, Phenylamine, Benzenamine, Aniline |
| Molecular Weight (g/mol) | 93.13 |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.