CAS RN 612-64-6
CAS RN 612-64-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso-N-ethylaniline, TRC
CAS: 612-64-6 Molecular Formula: C8 H10 N2 O Molecular Weight (g/mol): 150.18 Synonym: Aniline, N-ethyl-N-nitroso- (6CI,7CI,8CI),N-Ethyl-N-nitrosobenzenamine,N-Ethyl-N-nitrosoaniline,N-Ethyl-N-phenylnitrosamine,N-Nitroso-N-ethylaniline,N-Nitrosoethylphenylamine,NSC 405070,N-Nitroso-N-ethyl-N-phenylamine,NEPhA IUPAC Name: N-ethyl-N-phenylnitrous amide SMILES: CCN(N=O)c1ccccc1