accessibility menu, dialog, popup

UserName

CAS RN 60657-70-7

CH3HOH3CH3C

CAS RN 60657-70-7

IUPAC Name: 2-methyloct-5-yn-4-ol
Synonyms: 2-methyloct-5-yn-4-ol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: YBGQRUFRRPFNKW-UHFFFAOYNA-N
SMILES: CCC#CC(O)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results