missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-octyn-4-ol, 97%, Thermo Scientific™

Product Code. 11323276
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323276 25 g 25g
11313276 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323276 Supplier Thermo Scientific Alfa Aesar Supplier No. L12625.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60657-70-7
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00041690
InChI Key YBGQRUFRRPFNKW-UHFFFAOYNA-N
Synonym 2-methyl-5-octyn-4-ol, 5-octyn-4-ol, 2-methyl, acmc-20anql, 2-methyl-5-octyne-4-ol
PubChem CID 143720
IUPAC Name 2-methyloct-5-yn-4-ol
SMILES CCC#CC(O)CC(C)C

Specifications

Density 0.855
Flash Point 78°C (172°F)
Refractive Index 1.449
Quantity 25 g
Beilstein 8899472
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-octyn-4-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.