accessibility menu, dialog, popup

UserName

CAS RN 604-53-5

IUPAC Name: 1,1'-binaphthalene
Synonyms: 1,1'-binaphthalene
Molecular Weight (g/mol): 254.33
Molecular Formula: C20H14
InChi Key: ZDZHCHYQNPQSGG-UHFFFAOYSA-N
SMILES: C1=CC2=CC=CC(C3=CC=CC4=CC=CC=C34)=C2C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results