accessibility menu, dialog, popup

UserName

H3CNCH3(S)(S)(S)OHOH(S)CH3HOO(S)OHOH2NOHO(Z)

CAS RN 60-54-8

IUPAC Name: (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione
Synonyms: tetracycline
Molecular Weight (g/mol): 444.44
Molecular Formula: C22H24N2O8
InChi Key: JYHCQVWYCGHXGP-BPPSBWQWSA-N
SMILES: CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Color 
  • (5)
Physical Form 
  • (1)
  • (4)
Type 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)