accessibility menu, dialog, popup

UserName

CH3OHO(Z)(Z)

CAS RN 60-33-3

IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid
Synonyms: linoleic pure linoleic acid acide linoleique oils, grape oils, grape seed emersol 315 emersol 310 polylin no. 515 acide cis-linoleique grape seed oil
Molecular Weight (g/mol): 280.45
Molecular Formula: C18H32O2
InChi Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (11)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (5)
  • (1)
  • (5)
Physical Form 
  • (9)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (5)
  • (6)
  • (3)
  • (2)
  • (2)
  • (11)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (5)
  • (9)
  • (2)
  • (2)
  • (5)