missing translation for 'onlineSavingsMsg'
Learn More

Linoleic acid, 99.45%, MP Biomedicals™

Product Code. 11441901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441901 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441901 Supplier MP Biomedicals Supplier No. 0219469505
  • ONLINE EXCLUSIVE PRICE
    139.00 EUR
    132.30 EUR / 5g
    Save 6.70 EUR
    4% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 60-33-3
Molecular Formula C18H32O2
Quantity 5 g
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(O)=O
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
Molecular Weight (g/mol) 280.45
Formula Weight 280.44
Physical Form Liquid
Chemical Name or Material Linoleic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.