accessibility menu, dialog, popup

UserName

CH3(R)NH2(S)CH3

CAS RN 59480-92-1

IUPAC Name: (2R,5S)-2,5-dimethyl-2,5-dihydro-1H-pyrrol-1-ium
Synonyms: (2R,5S)-2,5-dimethyl-2,5-dihydro-1H-pyrrol-1-ium
Molecular Weight (g/mol): 98.17
Molecular Formula: C6H12N
InChi Key: TXQDHQBSNAJSHQ-OLQVQODUSA-O
SMILES: C[C@H]1[NH2+][C@@H](C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results