accessibility menu, dialog, popup

UserName

H3COO(S)HNOOCH3CH3CH3OOCH3

CAS RN 59279-60-6

IUPAC Name: 1,5-dimethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate
Synonyms: 1,5-dimethyl (2S)-2-[(tert-butoxycarbonyl)amino]pentanedioate
Molecular Weight (g/mol): 275.3
Molecular Formula: C12H21NO6
InChi Key: QNSPKWUAZQIIGZ-QMMMGPOBSA-N
SMILES: COC(=O)CC[C@H](NC(=O)OC(C)(C)C)C(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results