CAS RN 5910-87-2
CAS RN 5910-87-2
IUPAC Name:
(2E,4E)-nona-2,4-dienal
Synonyms:
trans, trans-nona-2,4-dienal
Molecular Weight (g/mol):
138.21
Molecular Formula:
C9H14O
InChi Key:
ZHHYXNZJDGDGPJ-BSWSSELBSA-N
SMILES:
CCCC\C=C\C=C\C=O
1
–
3
of
3
results
1
trans,trans-2,4-Nonadienal (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
trans,trans-2,4-Nonadienal (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
trans,trans-2,4-Nonadienal (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.