Learn More
trans,trans-2,4-Nonadienal (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | trans,trans-2,4-Nonadienal |
| CAS | 5910-87-2 |
| Molecular Formula | C9H14O |
| Quantity | 250 g |
| Synonym | (2E,4E)-2,4-Nonadienal, (E,E)-2,4-Nonadienal, (2E,4E)-2,4-Nonadienal, (2E,4E)-Nona-2,4-dienal, (2E,4E)-Nonadienal, (E,E)-2,4-Nonadien-1-al, trans-2,trans-4-Nonadienal |
| InChI Formula | InChI=1S/C9H14O/c1-2-3-4-5-6-7-8-9-10/h5-9H,2-4H2,1H3/b6-5+,8-7+ |
| SMILES | CCCC\C=C\C=C\C=O |
| IUPAC Name | (2E,4E)-nona-2,4-dienal |
| Molecular Weight (g/mol) | 138.21 |
| Formula Weight | 138.1045 |