accessibility menu, dialog, popup

UserName

CH3SOOOCH3NF

CAS RN 58086-67-2

IUPAC Name: 2-fluoro-1-methylpyridin-1-ium 4-methylbenzene-1-sulfonate
Synonyms: 2-fluoro-1-methylpyridin-1-ium tosylate
Molecular Weight (g/mol): 283.32
Molecular Formula: C13H14FNO3S
InChi Key: HQWDKLAIDBOLFE-UHFFFAOYSA-M
SMILES: C[N+]1=CC=CC=C1F.CC1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results