Learn More
2-Fluoro-1-methylpyridinium p-Toluenesulphonate (Technical Grade), TRC
Product Code. 30400339
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400339 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30400339 Supplier Toronto Research Chemicals Supplier No. TRCF598255500MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Fluoro-1-methylpyridinium p-Toluenesulphonate (Technical Grade)
CAS 58086-67-2
Molecular Formula C13H14FNO3S
Quantity 500 mg
Synonym 2-Fluoro-1-methylpyridinium Tosylate, 2-Fluoro-N-methylpyridinium Tosylate
InChI Formula InChI=1S/C7H8O3S.C6H7FN/c1-6-2-4-7(5-3-6)11(8,9)10;1-8-5-3-2-4-6(8)7/h2-5H,1H3,(H,8,9,10);2-5H,1H3/q;+1/p-1
SMILES Cc1ccc(cc1)S(=O)(=O)[O-].C[n+]2ccccc2F
IUPAC Name 2-fluoro-1-methylpyridin-1-ium;4-methylbenzenesulfonate
Molecular Weight (g/mol) 283.32
Formula Weight 283.0678