accessibility menu, dialog, popup

UserName

CAS RN 58-64-0

NH2NNNNOOPHOOOPOHHOOHOHO

CAS RN 58-64-0

IUPAC Name: [({[5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
Synonyms: adenosine diphosphate
Molecular Weight (g/mol): 427.2
Molecular Formula: C10H15N5O10P2
InChi Key: XTWYTFMLZFPYCI-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3OC(COP(O)(=O)OP(O)(O)=O)C(O)C3O)C2=NC=N1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)