accessibility menu, dialog, popup

UserName

CH3ONH2S

CAS RN 57774-79-5

IUPAC Name: 4-ethoxybenzene-1-carbothioamide
Synonyms: 4-ethoxybenzenecarbothioamide
Molecular Weight (g/mol): 181.25
Molecular Formula: C9H11NOS
InChi Key: AREJPGXIKMXBIQ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results