accessibility menu, dialog, popup

UserName

CAS RN 574-93-6

IUPAC Name: 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1,3,5,7,9,11,13,15,17,19,21(38),22,24,26,28,30(37),31,33,35-nonadecaene
Synonyms: phthalocyanine
Molecular Weight (g/mol): 514.55
Molecular Formula: C32H18N8
InChi Key: IEQIEDJGQAUEQZ-UHFFFAOYSA-N
SMILES: N1C2=NC3=C4C=CC=CC4=C(N3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)