missing translation for 'onlineSavingsMsg'
Learn More

Phthalocyanine, MP Biomedicals

Product Code. 11453555 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453555 10 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453555 Supplier MP Biomedicals Supplier No. 0521670005
  • ONLINE EXCLUSIVE PRICE
    113.00 EUR
    111.30 EUR / 5g
    Save 1.70 EUR
    1% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 574-93-6
Molecular Formula C32H18N8
Quantity 10 g
Synonym phthalocyanine, pigment blue 16, 29h,31h-phthalocyanine, phthalocyanin, heliogen blue g, polymon blue g, lionol blue kw, irgazin blue 3gt, ftalocianina, ci pigment blue 16
InChI Key IEQIEDJGQAUEQZ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3
Molecular Weight (g/mol) 514.552
PubChem CID 5282330
ChEBI CHEBI:34921
Formula Weight 514.55
Chemical Name or Material Phthalocyanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.