CAS RN 5719-85-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Tolbutamide, TRC
CAS: 5719-85-7 Molecular Formula: C12 H18 N2 O4 S Molecular Weight (g/mol): 286.35 Synonym: 4-Hydroxytolbutamide,N-[(Butylamino)carbonyl]-4-(hydroxymethyl)benzenesulfonamide (ACI),Urea, 1-butyl-3-[(α-hydroxy-p-tolyl)sulfonyl]- (6CI, 7CI, 8CI),1-Butyl-3-(p-hydroxymethylphenyl)sulfonylurea,1-Butyl-3-[4-(hydroxymethyl)phenyl]sulfonylurea,Hydroxymethylcarbutamide,Hydroxymethyltolbutamide,Hydroxytolbutamide IUPAC Name: 1-butyl-3-[4-(hydroxymethyl)phenyl]sulfonylurea SMILES: CCCCNC(=O)NS(=O)(=O)c1ccc(CO)cc1