accessibility menu, dialog, popup

UserName

CH3NClCl

CAS RN 56961-78-5

IUPAC Name: 2,4-dichloro-5-methylpyridine
Synonyms: 2,4-dichloro-5-methylpyridine
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: DUPJNQNLAKJWKU-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results