CAS RN 568544-03-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GSTO1-IN-1, MedChemExpress
MedChemExpress GSTO1-IN-1 is a potent glutathione S-transferase omega 1 (GSTO1) inhibitor with an IC50 of 31 nM.