missing translation for 'onlineSavingsMsg'
Learn More

GSTO1-IN-1, MedChemExpress

Product Code. 16465356
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465356 100 mg 100mg
16460797 50 mg 50mg
16454488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465356 Supplier Medchem Express Supplier No. HY111530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSTO1-IN-1 is a potent glutathione S-transferase omega 1 (GSTO1) inhibitor with an IC50 of 31 nM.

GSTO1-IN-1 is a potent glutathione S-transferase omega 1 (GSTO1) inhibitor with an IC50 of 31 nM.

Specifications

Chemical Name or Material GSTO1-IN-1
CAS 568544-03-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H12Cl2N2O3S
Quantity 100 mg
Solubility Information DMSO : ≥ 300 mg/mL (964.07 mM)
SMILES O=C(NC1=CC=C(Cl)C(S(=O)(N(C)C)=O)=C1)CCl
Molecular Weight (g/mol) 311.18
Formula Weight 311.18
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.