accessibility menu, dialog, popup

UserName

CAS RN 56771-29-0

IUPAC Name: (pent-4-en-1-yl)triphenylphosphanium bromide
Synonyms: pent-4-en-1-yltriphenylphosphanium bromide
Molecular Weight (g/mol): 411.32
Molecular Formula: C23H24BrP
InChi Key: KANICXDUFCDRDS-UHFFFAOYSA-M
SMILES: [Br-].C=CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results