CAS RN 55779-18-5
IUPAC Name:
9-[(2-chloro-6-fluorophenyl)methyl]-9H-purin-6-amine
Synonyms:
arprinocid arprinocida arprinocide arprinocidum 阿普西特 C12H9ClFN5 arpocox
Molecular Weight (g/mol):
277.69
Molecular Formula:
C12H9ClFN5
InChi Key:
NAPNOSFRRMHNBJ-UHFFFAOYSA-N
SMILES:
NC1=C2N=CN(CC3=C(F)C=CC=C3Cl)C2=NC=N1
1
–
3
of
3
results
1
Arprinocid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Arprinocid, TRC
CAS: 55779-18-5 Molecular Formula: C12 H9 Cl F N5 Molecular Weight (g/mol): 277.68 Synonym: 9-[(2-Chloro-6-fluorophenyl)methyl]-9H-purin-6-amine,9-[(2-Chloro-6-fluorophenyl)methyl]-9H-purin-6-amine,6-Amino-9-(2-chloro-6-fluorobenzyl)purine,Arpocox,Arprinocid,MK 302 IUPAC Name: 9-[(2-chloro-6-fluorophenyl)methyl]purin-6-amine SMILES: Nc1ncnc2c1ncn2Cc3c(F)cccc3Cl