Learn More
Arprinocid, Mikromol™
Description
Arprinocid
Specifications
Specifications
| Chemical Name or Material | Arprinocid |
| CAS | 55779-18-5 |
| Analyte or Component Names | Arprinocid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H9 Cl F N5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 9-[(2-Chloro-6-fluorophenyl)methyl]-9H-purin-6-amine, 6-Amino-9-(2-chloro-6-fluorobenzyl)purine, Arpocox, Arprinocid, MK 302 |
| InChI Formula | InChI=1S/C12H9ClFN5/c13-8-2-1-3-9(14)7(8)4-19-6-18-10-11(15)16-5-17-12(10)19/h1-3,5-6H,4H2,(H2,15,16,17) |
| SMILES | Nc1ncnc2c1ncn2Cc3c(F)cccc3Cl |
| IUPAC Name | 9-[(2-chloro-6-fluorophenyl)methyl]purin-6-amine |
| Show More |