accessibility menu, dialog, popup

UserName

OHOHO

CAS RN 552-63-6

IUPAC Name: 3-hydroxy-2-phenylpropanoic acid
Synonyms: (+-)-tropic acid tropic acid, (R)- tropic acid, (S)- (s)-tropic acid tropic acid, (s)-isomer (R)-isomer tropic acid 2-phenyl-hydracrylic acid tropic acid, (r)-isomer α-phenylhydracrylic acid (+-)-isomer tropic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: JACRWUWPXAESPB-UHFFFAOYNA-N
SMILES: OCC(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)