accessibility menu, dialog, popup

UserName

CAS RN 55179-31-2

H3CCH3H3COHONNN

CAS RN 55179-31-2

IUPAC Name: 1-{[1,1'-biphenyl]-4-yloxy}-3,3-dimethyl-1-(1H-1,2,4-triazol-1-yl)butan-2-ol
Synonyms: bitertanol bitertanol stereoisomer A sibutol biloxazol baymat-spray baycor baycor 25 WP
Molecular Weight (g/mol): 337.42
Molecular Formula: C20H23N3O2
InChi Key: VGPIBGGRCVEHQZ-UHFFFAOYNA-N
SMILES: CC(C)(C)C(O)C(OC1=CC=C(C=C1)C1=CC=CC=C1)N1C=NC=N1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Concentration 
  • (3)
  • (3)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)