missing translation for 'onlineSavingsMsg'
Learn More

Bitertanol, TRC

Product Code. 30336588
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336588 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336588 Supplier Toronto Research Chemicals Supplier No. TRCB3865001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bitertanol
CAS 55179-31-2
Molecular Formula C20 H23 N3 O2
Quantity 1 g
Synonym β-([1,1'-Biphenyl]-4-yloxy)-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol, Baycor, Baycor 25WP, Baycor DC 300, Biloxazol, Bitertanol, KWG 0599
InChI Formula InChI=1S/C20H23N3O2/c1-20(2,3)18(24)19(23-14-21-13-22-23)25-17-11-9-16(10-12-17)15-7-5-4-6-8-15/h4-14,18-19,24H,1-3H3
SMILES CC(C)(C)C(O)C(Oc1ccc(cc1)c2ccccc2)n3cncn3
IUPAC Name 3,3-dimethyl-1-(4-phenylphenoxy)-1-(1,2,4-triazol-1-yl)butan-2-ol
Molecular Weight (g/mol) 337.42
Formula Weight 337.179
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.