CAS RN 548-93-6
CAS RN 548-93-6
IUPAC Name:
2-amino-3-hydroxybenzoic acid
Synonyms:
3-hydroxyanthranilic acid 3-hydroxy-anthranilsaeure 3-ohaa 3-hydroxyantranilic acid 3 hydroxyanthranilic acid acid, 3-hydroxyanthranilic 3-hydroxanthranilate 3-hydroxy-anthranilate 3-OH-anthranilic acid
Molecular Weight (g/mol):
153.14
Molecular Formula:
C7H7NO3
InChi Key:
WJXSWCUQABXPFS-UHFFFAOYSA-N
SMILES:
NC1=C(O)C=CC=C1C(O)=O
1
–
2
of
2
results
2
3-Hydroxyanthranilic acid, MedChemExpress
MedChemExpress 3-Hydroxyanthranilic acid is a tryptophan metabolite in the kynurenine pathway.