Learn More
3-Hydroxyanthranilic acid, MedChemExpress
Product Code. 16489306
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16489306
Brand: Medchem Express HYW001171
MedChemExpress 3-Hydroxyanthranilic acid is a tryptophan metabolite in the kynurenine pathway.
3-Hydroxyanthranilic acid is a tryptophan metabolite in the kynurenine pathway.
Specifications
Chemical Name or Material 3-Hydroxyanthranilic acid
CAS 548-93-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H7NO3
Solubility Information DMSO : 50 mg/mL (326.50 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC(O)=C1N
Molecular Weight (g/mol) 153.14
Quantity 100 mg
Formula Weight 153.14
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism