accessibility menu, dialog, popup

UserName

CAS RN 548-04-9

CH3OHOOHHOHOHOOHOCH3

CAS RN 548-04-9

IUPAC Name: 7,11,13,16,18,22-hexahydroxy-5,24-dimethyloctacyclo[13.11.1.1²,¹⁰.0³,⁸.0⁴,²⁵.0¹⁹,²⁷.0²¹,²⁶.0¹⁴,²⁸]octacosa-1(27),2(28),3,5,7,10,12,14,16,18,21,23,25-tridecaene-9,20-dione
Synonyms: 7,11,13,16,18,22-hexahydroxy-5,24-dimethyloctacyclo[13.11.1.1²,¹⁰.0³,⁸.0⁴,²⁵.0¹⁹,²⁷.0²¹,²⁶.0¹⁴,²⁸]octacosa-1(27),2(28),3,5,7,10,12,14,16,18,21,23,25-tridecaene-9,20-dione mono-(123I)iodohypericin hypericin hypericine hypericum red hyperizin (component of) hypericum spp (st. john wort) (component of) hypericum spp (st. john's wort) hipericina mono-(123i)iodohypericin
Molecular Weight (g/mol): 504.45
Molecular Formula: C30H16O8
InChi Key: BTXNYTINYBABQR-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C2C(=O)C3=C(O)C=C(O)C4=C5C(O)=CC(O)=C6C(=O)C7=C(O)C=C(C)C8=C7C(=C56)C(C2=C18)=C34
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)