accessibility menu, dialog, popup

UserName

CAS RN 54796-47-3

CH3SOOH3CBr

CAS RN 54796-47-3

IUPAC Name: methyl 5-bromo-4-methylthiophene-2-carboxylate
Synonyms: methyl 5-bromo-4-methylthiophene-2-carboxylate
Molecular Weight (g/mol): 235.1
Molecular Formula: C7H7BrO2S
InChi Key: RJWZFHRMQRWEIH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C)=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results