CAS RN 545338-89-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl] Sumatriptan (>90%), TRC
CAS: 545338-89-4 Molecular Formula: C27 H37 N5 O2 S Molecular Weight (g/mol): 495.68 Synonym: 1H-Indole-5-methanesulfonamide, 3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-N-methyl-,[3-[2-(Dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide,Sumatriptan Succinate Imp. A (EP) IUPAC Name: 1-[3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide SMILES: CNS(=O)(=O)Cc1ccc2[nH]c(Cc3ccc4[nH]cc(CCN(C)C)c4c3)c(CCN(C)C)c2c1