accessibility menu, dialog, popup

UserName

CAS RN 5419-02-3

CH3NHH3CHO

CAS RN 5419-02-3

IUPAC Name: [(2-hydroxynaphthalen-1-yl)methyl]dimethylazanium
Synonyms: [(2-hydroxynaphthalen-1-yl)methyl]dimethylazanium
Molecular Weight (g/mol): 202.28
Molecular Formula: C13H16NO
InChi Key: GOHHBFSPKLRUNF-UHFFFAOYSA-O
SMILES: C[NH+](C)CC1=C2C=CC=CC2=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results