accessibility menu, dialog, popup

UserName

ZnOSOOFFFOSOOFFF

CAS RN 54010-75-2

IUPAC Name: zinc(2+) ditrifluoromethanesulfonate
Synonyms: zinc trifluoromethanesulfonate
Molecular Weight (g/mol): 363.51
Molecular Formula: C2F6O6S2Zn
InChi Key: CITILBVTAYEWKR-UHFFFAOYSA-L
SMILES: [Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)