missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) Trifluoromethanesulphonate, TRC

Product Code. 30457232
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457232 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457232 Supplier Toronto Research Chemicals Supplier No. TRCZ43968350MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc(II) Trifluoromethanesulphonate
CAS 54010-75-2
Molecular Formula C F3 O3 S . Zn
Quantity 50 mg
InChI Formula InChI=1S/CHF3O3S.Zn/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+2/p-1
SMILES [Zn+2].[O-]S(=O)(=O)C(F)(F)F
IUPAC Name zinc;trifluoromethanesulfonate
Molecular Weight (g/mol) 214.478
Formula Weight 212.881
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.