accessibility menu, dialog, popup

UserName

H3COOCH3H3CCH3H2NHCl

CAS RN 53956-05-1

IUPAC Name: tert-butyl 2-aminobutanoate hydrochloride
Synonyms: tert-butyl 2-aminobutanoate hydrochloride
Molecular Weight (g/mol): 195.69
Molecular Formula: C8H18ClNO2
InChi Key: OEPKETQBSXWOBJ-UHFFFAOYNA-N
SMILES: Cl.CCC(N)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results