accessibility menu, dialog, popup

UserName

CAS RN 533-50-6

HO(S)OHOOH

CAS RN 533-50-6

IUPAC Name: 1,3,4-trihydroxybutan-2-one
Synonyms: erythrulose
Molecular Weight (g/mol): 120.1
Molecular Formula: C4H8O4
InChi Key: UQPHVQVXLPRNCX-UHFFFAOYNA-N
SMILES: OCC(O)C(=O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results