accessibility menu, dialog, popup

UserName

CAS RN 53188-07-1

IUPAC Name: 6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Synonyms: trolox trolox C
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: GLEVLJDDWXEYCO-UHFFFAOYNA-N
SMILES: CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)