accessibility menu, dialog, popup

UserName

H3COS

CAS RN 53119-25-8

IUPAC Name: 1-(thiophen-2-yl)pentan-1-one
Synonyms: 1-(thiophen-2-yl)pentan-1-one
Molecular Weight (g/mol): 168.25
Molecular Formula: C9H12OS
InChi Key: KJMPRCVRMAHTBR-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results