CAS RN 53-70-3
IUPAC Name:
pentacyclo[12.8.0.0³,¹².0⁴,⁹.0¹⁵,²⁰]docosa-1,3(12),4,6,8,10,13,15,17,19,21-undecaene
Synonyms:
dibenz(A,H)anthracene DBA dibenzo(a,h)anthracene 1,2,5,6-dibenzanthraceen 1,2,5,6-dibenzanthracene DB(a,h)A 1,2:5,6-benzanthracene ccris 208 1,2:5,6-dibenzanthracene rcra waste number U063
Molecular Weight (g/mol):
278.35
Molecular Formula:
C22H14
InChi Key:
LHRCREOYAASXPZ-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(C=CC3=CC4=C(C=CC5=CC=CC=C45)C=C23)=C1
1
–
8
of
8
results
2
Dibenz[a,h]anthracene, TRC
CAS: 53-70-3 Molecular Formula: C22 H14 Molecular Weight (g/mol): 278.35 Synonym: 1,2:5,6-Benzanthracene,1,2:5,6-Dibenz[a]anthracene,1,2:5,6-Dibenzanthracen,1,2:5,6-Dibenzanthracene,1,2:5,6-Dibenzoanthracene,DBA,Dibenzo[a,h]anthracene,NSC 22433 IUPAC Name: naphtho[1,2-b]phenanthrene SMILES: c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23