Learn More
Metabolite Dibenzo(a,h)anthracene in DCM 10μg/mL, Fisher Chemical™
Product Code. 12935294 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12935294 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12935294 Supplier Fisher Chemical Supplier No. P816950
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule(s)
CAS 53-70-3
Molecular Formula C22H14
Concentration 10 μg/mL
InChI Key LHRCREOYAASXPZ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=CC4=C(C=CC5=CC=CC=C54)C=C32
IUPAC Name naphtho[1,2-b]phenanthrene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 278.354
PubChem CID 5889
Quantity 10 mL
ChEBI CHEBI:35299
Chemical Name or Material Dibenzo(a,h)anthracene