CAS RN 519-32-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,3,9-Trimethyl-3,9-dihydro-1H-purine-2,6-dione (Isocaffeine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Isocaffeine, TRC
CAS: 519-32-4 Molecular Formula: C8 H10 N4 O2 Molecular Weight (g/mol): 194.19 Synonym: 1,3,9-Trimethyl-3,9-dihydro-1H-purine-2,6-dione,Isocaffeine,Caffeine Imp. C (EP) IUPAC Name: 1,3,9-trimethylpurine-2,6-dione SMILES: CN1C(=O)N(C)c2c(ncn2C)C1=O