accessibility menu, dialog, popup

UserName

CAS RN 51609-06-4

H3CH3CCH3NH

CAS RN 51609-06-4

IUPAC Name: N-tert-butylcyclohexanamine
Synonyms: N-tert-butylcyclohexanamine
Molecular Weight (g/mol): 155.29
Molecular Formula: C10H21N
InChi Key: MTEWAFVECQBILW-UHFFFAOYSA-N
SMILES: CC(C)(C)NC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results