accessibility menu, dialog, popup

UserName

INOO

CAS RN 516-12-1

IUPAC Name: 1-iodopyrrolidine-2,5-dione
Synonyms: N-iodosuccinimide succiniodimide NIS
Molecular Weight (g/mol): 224.99
Molecular Formula: C4H4INO2
InChi Key: LQZMLBORDGWNPD-UHFFFAOYSA-N
SMILES: IN1C(=O)CCC1=O
Quantity 
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)